4-Amino-3-iodobenzonitrile ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
4-Cyano-2-iodoaniline
Palladium-catalyzed carbonylation of iodoanilines provides ketoamides in the presence of primary and secondary amines.Reactant involved in: 1.Hydroamination-double hydroarylation for synthesis of fuzed carbazoles. 2. Cycloisomerizations of aromatic homo- and bis-homopropargylic amine / amide intermediates. 3.Asymmetric organocatalytic condensation and cycloaddition. 4. Regioselective aryl C-H bond functionalization. 5.Synthesis of [(triisopropylsilyl)ethynyl]indole derivatives. 6.Domino condensation / S-arylation / heterocyclization reactions. 7.Palladium-catalyzed carbonylation of iodoanilines provides ketoamides in the presence of primary and secondary amines.
Formula:
C₇H₅IN₂ MW: 244.03 g/mol Melting Pt: 112-115°C |
MDL Number:
MFCD04039964 CAS Number: 33348-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...