1-​Amino-​1-​deoxy-​β-​D-​galactose ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: β-D-Galactosylamine, 1-Amino-1-deoxy-β-D-galactose

A120845-1G A120845-250MG
ALA120845-1GEA 1729.86 USD
ALA120845-1G ALA120845250MG
1-​Amino-​1-​deoxy-​β-​D-​galactose ≥98%
1-​Amino-​1-​deoxy-​β-​D-​galactose

β-D-Galactosylamine, a galactose analogue, is used as a competitive inhibitor to help isolate, purify, identify, differentiate and characterize β-D-galactosidase(s) and galactose oxidase(s).

Formula: C₆H₁₃NO₅
MW: 179.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00047541
CAS Number: 74867-91-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3db04c742d94ee2510e642e693ef136934819208&vid=3db04c742d94ee2510e642e693ef136934819208&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR