2-Amino-3-bromo-6-methylpyridine ≥98%

Supplier: Aladdin Scientific
Warning

A120782-1G A120782-25G A120782-5G
ALA120782-1GEA 33.16 USD
ALA120782-1G ALA120782-25G ALA120782-5G
2-Amino-3-bromo-6-methylpyridine ≥98%
2-Amino-3-bromo-6-methylpyridine
Formula: C₆H₇BrN₂
Boiling Pt: 238.5±35.0 °C
Melting Pt: 77-79°C
Density: 1.5672 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD03095215
CAS Number: 126325-46-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=333ecc172600a35793f9c6d0752a24dfc2889d64&vid=333ecc172600a35793f9c6d0752a24dfc2889d64&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR