2,3,4,6-Tetra-O-acetyl-α-D-galactopyranosyl bromide ≥93% stabilised

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Acetobromo-α-D-galactose, (2R,3S,4S,5R,6R)-2-(acetoxymethyl)-6-bromotetrahydro-2H-pyran-3,4,5-triyl triacetate, Tetra-O-acetyl-α-D-galactopyranosyl bromide, Acetobromo-a-D-galactose

Stabilization: Stabilised with calcium carbonate 2%

A120774-1G A120774-25G A120774-5G
ALA120774-1GEA 9.41 USD
ALA120774-1G ALA120774-25G ALA120774-5G
2,3,4,6-Tetra-O-acetyl-α-D-galactopyranosyl bromide ≥93% stabilised
2,3,4,6-Tetra-O-acetyl-alpha-D-galactopyranosyl bromide

2,3,4,6-Tetra-O-acetyl-α-D-galactopyranosyl bromide is used as an organic condensation reagent for chemistries such as N- and S- galactosylation reactions and the synthesis of spacer-equipped phosphorylated di-, tri- and tetrasaccharide fragments of the O-specific polysaccharides.

Formula: C₁₄H₁₉BrO₉
Storage Temperature: Ambient
MDL Number: MFCD00063686
CAS Number: 3068-32-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=300f1ad419de359a973aa0532d8d042416678994&vid=300f1ad419de359a973aa0532d8d042416678994&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR