2-Aminopyridine-6-methanol ≥97%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
(6-Aminopyridin-2-yl)methanol
A120737-1G
A120737-250MG
A120737-5G
ALA120737-1GEA
163.31
USD
ALA120737-1G
ALA120737250MG
ALA120737-5G
2-Aminopyridine-6-methanol ≥97%
2-Aminopyridine-6-methanol
Formula:
C₆H₈N₂O MW: 124.14 g/mol Melting Pt: 87-92°C |
MDL Number:
MFCD09863097 CAS Number: 79651-64-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...