5-Amino-2-fluorobenzoic acid ≥99%

Supplier: ALADDIN SCIENTIFIC
Warning

A120665-1G A120665-500MG A120665-5G
ALA120665-1GEA 14.16 USD
ALA120665-1G ALA120665500MG ALA120665-5G
5-Amino-2-fluorobenzoic acid ≥99%
5-Amino-2-fluorobenzoic acid
Formula: C₇H₆FNO₂
Melting Pt: 190-192°C
MDL Number: MFCD00077449
CAS Number: 56741-33-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53e952ad059f19bdb8560bf163bdfd472b6e1104&vid=53e952ad059f19bdb8560bf163bdfd472b6e1104&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR