6-Aminopicolinic acid ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 6-Aminopyridine-2-carboxylic acid

A120659-100G A120659-1G A120659-25G A120659-5G
ALA120659-100GEA 2461.36 USD
ALA120659-100G ALA120659-1G ALA120659-25G ALA120659-5G
6-Aminopicolinic acid ≥98%
6-Aminopicolinic acid
Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Melting Pt: 295°C
MDL Number: MFCD00233711
CAS Number: 23628-31-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e975a733fcde8fee2df23668669f5c749daf23b1&vid=e975a733fcde8fee2df23668669f5c749daf23b1&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR