2-Aminophenylboronic acid pinacol ester ≥97%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 2-Aminobenzeneboronic acid pinacol ester
A120514-100G
A120514-1G
A120514-25G
A120514-5G
ALA120514-100GEA
2803.36
USD
ALA120514-100G
ALA120514-1G
ALA120514-25G
ALA120514-5G
2-Aminophenylboronic acid pinacol ester ≥97%
2-Aminophenylboronic acid pinacol ester
Reactant involved in synthesis of: Indolo[3,4-cd][1]benzazepines via Pictet-Spengler-type cyclizations Antimicrobial amphiphilic aryl peptideomimetics Pyridoquinazolines and benzo[h][1,6]naphthyridines via intramolecular electrophilic substitution reactions Thienopyridine derivatives Dihydroquinolones Methacrylamidophenylboronic acids
Formula:
C₁₂H₁₈BNO₂ Boiling Pt: 125 °C Melting Pt: 65-70°C |
MDL Number:
MFCD02179448 CAS Number: 191171-55-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...