4-Aminophenylboronic acid pinacol ester ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-Aminobenzeneboronic acid pinacol ester

A120513-1G A120513-25G A120513-5G
ALA120513-1GEA 52.16 USD
ALA120513-1G ALA120513-25G ALA120513-5G
4-Aminophenylboronic acid pinacol ester ≥98%
4-Aminophenylboronic acid pinacol ester

Reagent used for ? Suzuki-Miyaura cross-coupling reactions. ? Mercury(II) detection by fluorometry with new fluorogenic indicators based on through-bond energy transfer from pentaquinone to rhodamine. ? Rhodium-catalyzed amination reactions. ? Palladium-catalyzed Suzuki cross-coupling to synthesize potential antitubercular and antimicrobial compounds.

Formula: C₁₂H₁₈BNO₂
MW: 219.09
Melting Pt: 166-170°C
Storage Temperature: Ambient
MDL Number: MFCD02093721
CAS Number: 214360-73-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5514b3929ff858f2fb97bdc0a47b38d4c523e580&vid=5514b3929ff858f2fb97bdc0a47b38d4c523e580&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR