2-Aminophenylboronic acid hydrochloride ≥95%
Supplier: Aladdin Scientific
|
Warning
|
A120509-1G
A120509-25G
A120509-5G
ALA120509-1GEA
19.86
USD
ALA120509-1G
ALA120509-25G
ALA120509-5G
2-Aminophenylboronic acid hydrochloride ≥95%
2-Aminophenylboronic acid hydrochloride
Reactant for: Preparation of small molecules with a trihydroxybenzene motif as pan-selectin antagonists and potential antiinflammatory agents Preparation of designed boronate ligands for glucose-selective holographic sensors
Formula:
C₆H₉BClNO₂ Melting Pt: 145-156°C Storage Temperature: Ambient |
MDL Number:
MFCD02258096 CAS Number: 863753-30-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...