2-Amino-4-nitrophenol sodium salt ≥80%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Amino-4-nitrophenol Sodium Salt, Sodium 2-amino-4-nitrophenolate, 2-Hydroxy-5-nitroaniline Sodium Salt

A120316-100G A120316-250G A120316-25G
ALA120316-100GEA 42.66 USD
ALA120316-100G ALA120316-250G ALA120316-25G
2-Amino-4-nitrophenol sodium salt ≥80%
2-Amino-4-nitrophenol sodium salt
Formula: C₆H₆N₂O₃
MW: 176.11 g/mol
MDL Number: MFCD00067428
CAS Number: 61702-43-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cec536a878e5218c6a4c0ff2e07f83c34096bc21&vid=cec536a878e5218c6a4c0ff2e07f83c34096bc21&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR