5-Amino-1-phenyl-1H-pyrazole-4-carboxylic acid ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A120240-1G A120240-5G
ALA120240-1GEA 74.96 USD
ALA120240-1G ALA120240-5G
5-Amino-1-phenyl-1H-pyrazole-4-carboxylic acid ≥97%
5-Amino-1-phenyl-1H-pyrazole-4-carboxylic acid
Formula: C₁₀H₉N₃O₂
MW: 203.2 g/mol
Melting Pt: 183-188°C
MDL Number: MFCD00220607
CAS Number: 51649-80-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e17bb7b53c2393f5c1bab8a51f7b1b7cd0669a41&vid=e17bb7b53c2393f5c1bab8a51f7b1b7cd0669a41&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR