Alfacalcidol ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: (1R,3S,Z)-5-((E)-2-((1R,3aS,7aR)-7a-Methyl-1-((R)-6-methylheptan-2-yl)hexahydro-1H-inden-4(2H)-ylidene)ethylidene)-4-methylenecyclohexane-1,3-diol, 1alpha-Hydroxyvitamin D3, 1alpha-Hydroxycholecalciferol

A120129-10MG A120129-50MG A120129-5MG
ALA120129-10MGEA 123.41 USD
ALA120129-10MG ALA120129-50MG ALA120129-5MG
Alfacalcidol ≥97%
Alfacalcidol
Formula: C₂₇H₄₄O₂
MW: 400.65 g/mol
Melting Pt: 136 °C
Storage Temperature: Freezer
MDL Number: MFCD00868328
CAS Number: 41294-56-8
EINECS: 255-297-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94410d7ecf6b3f62dc381457d82c3cc3bf5c081d&vid=94410d7ecf6b3f62dc381457d82c3cc3bf5c081d&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR