9-Anthraceneboronic acid ≥99% (contains varying amounts of Anhydride)
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Synonyms:
9-anthraceneboronic acid, Anthracene-9-boronic acid, anthracen-9-ylboronic acid
A120114-1G
A120114-250MG
A120114-5G
ALA120114-1GEA
24.61
USD
ALA120114-1G
ALA120114250MG
ALA120114-5G
9-Anthraceneboronic acid ≥99% (contains varying amounts of Anhydride)
9-Anthraceneboronic acid
Formula:
C₁₄H₁₁BO₂ MW: 222.05 g/mol Boiling Pt: 479.5°C Melting Pt: 203-250 ºC Flash Pt: 243.8ºC Storage Temperature: Ambient |
MDL Number:
MFCD03425925 CAS Number: 100622-34-2 UN: 2811 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...