2-Anthraceneboronic acid 97 (contains varying amounts of Anhydride)
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
Synonyms:
2-Anthraceneboronic acid, Anthracen-2-ylboronic acid, 2-Anthraceneboronic Acid
A120113-1G
A120113-250MG
ALA120113-1GEA
143.36
USD
ALA120113-1G
ALA120113250MG
2-Anthraceneboronic acid 97 (contains varying amounts of Anhydride)
2-Anthraceneboronic acid
Formula:
C₁₄H₁₁BO₂ MW: 222.05 g/mol |
MDL Number:
MFCD07784001 CAS Number: 141981-64-8 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...