3-Aminotetrahydrofuran ≥95%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: Tetrahydrofuran-3-amine

A120073-1G A120073-250MG A120073-5G
ALA120073-1GEA 131.96 USD
ALA120073-1G ALA120073250MG ALA120073-5G
3-Aminotetrahydrofuran ≥95%
3-Aminotetrahydrofuran
Formula: C₄H₉NO
MW: 87.12 g/mol
Boiling Pt: 73°C
Density: 1.012 g/cm³
Flash Pt: 48 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07778394
CAS Number: 88675-24-5
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99db267f52749f0d1e1b34a81be655f94c238714&vid=99db267f52749f0d1e1b34a81be655f94c238714&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR