4-Aminophenylboronic acid hydrochloride ≥97% (contains varying amounts of Anhydride)

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 4-Aminobenzeneboronic acid hydrochloride, (4-Aminophenyl)boronic acid hydrochloride

A120027-1G A120027-250MG A120027-500MG A120027-5G
ALA120027-1GEA 386.56 USD
ALA120027-1G ALA120027250MG ALA120027500MG ALA120027-5G
4-Aminophenylboronic acid hydrochloride ≥97% (contains varying amounts of Anhydride)
4-Aminophenylboronic acid hydrochloride

Preparation of chromen-4-ones as DNA-dependent protein kinase inhibitors.

Formula: C₆H₉BClNO₂
MW: 173.41 g/mol
Melting Pt: 125-132°C
Storage Temperature: Ambient
MDL Number: MFCD03001333
CAS Number: 80460-73-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a867c70c1b091eed26123405ec9e05a2b37d9109&vid=a867c70c1b091eed26123405ec9e05a2b37d9109&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR