Adapalene ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

A120006-1G A120006-250MG A120006-5G
ALA120006-1GEA 357.11 USD
ALA120006-1G ALA120006250MG ALA120006-5G
Adapalene ≥98%, Moligand™
Adapalene

Adapalene is a dual RAR and RXR agonist, used in the treatment of acne.Adapalene, a retinoic acid analogue, is a RARβ and RARγ agonist. Adapalene inhibits proliferation and induces apoptosis in colorectal cancer cells in vitro. Adapalene gel is an efficacious treatment for acne vulgaris.

Formula: C₂₈H₂₈O₃
Melting Pt: 301°C
Storage Temperature: Refrigerator
MDL Number: MFCD03106112
CAS Number: 106685-40-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a6344ad54afc6b45442429dd0a8c8d6cdd8c2b8&vid=0a6344ad54afc6b45442429dd0a8c8d6cdd8c2b8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR