3-Amino-1-propanesulphonic acid sodium salt ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A119944-100G A119944-1G A119944-25G A119944-5G
ALA119944-100GEA 1290.01 USD
ALA119944-100G ALA119944-1G ALA119944-25G ALA119944-5G
3-Amino-1-propanesulphonic acid sodium salt ≥98%
3-Amino-1-propanesulphonic acid sodium salt

GABAA receptor agonist.

Formula: C₃H₈NNaO₃S
Storage Temperature: Refrigerator
MDL Number: MFCD00064431
CAS Number: 81028-90-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fec6202b00f344e86acf2c60d501a484c8f7e79e&vid=fec6202b00f344e86acf2c60d501a484c8f7e79e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR