3-Fluoro-4-(trifluoromethyl)aniline ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: 3-Fluoro-4-trifluoromethylaniline, 4-Amino-alpha,alpha,alpha,2-tetrafluorotoluene, α,α,α,3-Tetrafluoro-p-toluidine, 4-Amino-2-fluorobenzotrifluoride, 4-Amino-α,α,α,2-tetrafluorotoluene

A119815-5G A119815-100G A119815-1G A119815-25G
ALA119815-5GEA 167.11 USD
ALA119815-5G ALA119815-100G ALA119815-1G ALA119815-25G
3-Fluoro-4-(trifluoromethyl)aniline ≥97%
3-Fluoro-4-(trifluoromethyl)aniline
Formula: C₇H₅F₄N
MW: 179.12 g/mol
Melting Pt: 54-58°C
Storage Temperature: Ambient
MDL Number: MFCD00190120
CAS Number: 69411-68-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8eda6070950149dfc1ffe665d29e6ede3bce3248&vid=8eda6070950149dfc1ffe665d29e6ede3bce3248&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR