4-Amino-3-(trifluoromethyl)benzonitrile ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: 2-Amino-5-cyanobenzotrifluoride

A119801-100G A119801-1G A119801-25G A119801-5G
ALA119801-100GEA 2734.01 USD
ALA119801-100G ALA119801-1G ALA119801-25G ALA119801-5G
4-Amino-3-(trifluoromethyl)benzonitrile ≥98%
4-Amino-3-(trifluoromethyl)benzonitrile
Formula: C₈H₅F₃N₂
MW: 186.14 g/mol
Melting Pt: 60-63°C
MDL Number: MFCD00275473
CAS Number: 327-74-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9eea22e052a8b65618a130f02f9b50f9e0e1419f&vid=9eea22e052a8b65618a130f02f9b50f9e0e1419f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR