(S)-2-Aminobutanamide hydrochloride ≥99%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: H-Abu-NH2 · HCl, (2S)-2-Aminobutanamide hydrochloride, (S)-(+)-2-Aminobutanamide hydrochloride, H-Abu-NH2 HCl

A119770-1G A119770-250MG A119770-25G A119770-5G
ALA119770-1GEA 19.86 USD
ALA119770-1G ALA119770250MG ALA119770-25G ALA119770-5G
(S)-2-Aminobutanamide hydrochloride ≥99%
(S)-2-Aminobutanamide hydrochloride
Formula: C₄H₁₁ClN₂O
MW: 138.60 g/mol
Melting Pt: 259-263°C
MDL Number: MFCD00136565
CAS Number: 7682-20-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f17f5f44fe19aab8d01b8bec049a64cc68aef6c0&vid=f17f5f44fe19aab8d01b8bec049a64cc68aef6c0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR