5-Amino-2-fluoropyridine ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A119632-10G A119632-1G A119632-25G A119632-5G
ALA119632-10GEA 102.51 USD
ALA119632-10G ALA119632-1G ALA119632-25G ALA119632-5G
5-Amino-2-fluoropyridine ≥98%
5-Amino-2-fluoropyridine
Formula: C₅H₅FN₂
Melting Pt: 88.0-92.0°C
Storage Temperature: Refrigerator
MDL Number: MFCD01632180
CAS Number: 1827-27-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3461517b70cb384d41801cb874345e00950a133a&vid=3461517b70cb384d41801cb874345e00950a133a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR