4-Amino-3-fluoropyridine ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A119629-1G A119629-5G
ALA119629-1GEA 56.91 USD
ALA119629-1G ALA119629-5G
4-Amino-3-fluoropyridine ≥97%
4-Amino-3-fluoropyridine
Formula: C₅H₅FN₂
Melting Pt: 78-82°C
Storage Temperature: Ambient
MDL Number: MFCD00234072
CAS Number: 2247-88-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfb5f02594c1001a15a6fa697802372a728dcb0d&vid=dfb5f02594c1001a15a6fa697802372a728dcb0d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR