3-Amino-2-fluoropyridine ≥95%
Supplier: ALADDIN SCIENTIFIC
|
Warning
|
A119619-1G
A119619-25G
A119619-5G
ALA119619-1GEA
65.46
USD
ALA119619-1G
ALA119619-25G
ALA119619-5G
3-Amino-2-fluoropyridine ≥95%
3-Amino-2-fluoropyridine
Formula:
C₅H₅FN₂ Density: 1.212 g/cm³ Flash Pt: >110 °C |
MDL Number:
MFCD03095248 CAS Number: 1597-33-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...