Antimony (III) sulfide ≥99.99% (metals basis), PrimorTrace™

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: Needle antimony, Antimony(III) sulphide, Amtimony sulphide, Diantimony trisulphide, Stibnite, Antimony glance, Antimony trisulphide, Antimonous sulphide, Antimony sulphide black

A119486-100G A119486-25G A119486-5G
ALA119486-100GEA 1051.56 USD
ALA119486-100G ALA119486-25G ALA119486-5G
Antimony (III) sulfide ≥99.99% (metals basis), PrimorTrace™
Antimony (III) sulphide
Formula: S₃Sb₂
MW: 339.72 g/mol
Melting Pt: 550°C
Density: 4.562 g/cm³
MDL Number: MFCD00011217
CAS Number: 1345-04-6
EINECS: 215-713-4
UN: 1549
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5371b6a432303f8abf88018d36829545296ff0d3&vid=5371b6a432303f8abf88018d36829545296ff0d3&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR