Antimony (III) sulphide ≥99.9% (metals basis)

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Needle antimony, Antimony(III) sulphide, Amtimony sulphide, Diantimony trisulphide, Stibnite, Antimony glance, Antimony trisulphide, Antimonous sulphide, Antimony sulphide black

A119485-10G A119485-250G A119485-50G
ALA119485-10GEA 66.41 USD
ALA119485-10G ALA119485-250G ALA119485-50G
Antimony (III) sulphide ≥99.9% (metals basis)
Antimony (III) sulphide
Formula: S₃Sb₂
MW: 339.72 g/mol
Boiling Pt: 1150
Melting Pt: 550
MDL Number: MFCD00011217
CAS Number: 1345-04-6
EINECS: 215-713-4
UN: 1549
ADR: 6.1 III
Merck Index: 12,00754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbe4a53c4ed51eaa3e6e8ed87207ba6e0bebcacf&vid=bbe4a53c4ed51eaa3e6e8ed87207ba6e0bebcacf&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR