Calcium ascorbate dihydrate ≥99%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Calcium (R)-5-((S)-1,2-dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one salt dihydrate, Vitamin C calcium salt, L-Ascorbic acid calcium salt dihydrate, Calcium L-ascorbate dihydrate

A119451-100G A119451-25G
ALA119451-100GEA 45.51 USD
ALA119451-100G ALA119451-25G
Calcium ascorbate dihydrate ≥99%
Calcium ascorbate dihydrate

Calcium L-ascorbate dihydrate, commonly called vitamin C calcium salt, is a biologically important antioxidant and a required coenzyme for various hydroxylation reactions and for collagen synthesis.A physiological antioxidant and coenzyme.

Formula: C₁₂H₁₄CaO₁₂·2H₂O
Melting Pt: 166°C
Flash Pt: 300 °C
MDL Number: MFCD00149216
CAS Number: 5743-28-2
EINECS: 227-261-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7c365b2f484505d7c66f427ad4f9e92ad364e56&vid=a7c365b2f484505d7c66f427ad4f9e92ad364e56&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR