Nickel aluminium alloy 48-50% Ni basis, powder 150 um

Supplier: ALADDIN SCIENTIFIC CORPORATION
Danger

Synonyms: Nickel-aluminum alloy, Raney®-Nickel alloy, Aluminum-nickel alloy

A119428-100G A119428-25G
ALA119428-100GEA 132.91 USD
ALA119428-100G ALA119428-25G
Nickel aluminium alloy 48-50% Ni basis, powder 150 um
Nickel aluminium alloy
Formula: AlNi
MW: 85.675 g/mol
MDL Number: MFCD00167392
CAS Number: 12635-27-7
UN: 3089
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2d3157c7df6f4a09ac2a2f08a5581883d20f48be&vid=2d3157c7df6f4a09ac2a2f08a5581883d20f48be&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR