2-Amino-6-bromopyridine ≥98%
Supplier: ALADDIN SCIENTIFIC CORPORATION
|
Warning
|
Synonyms:
6-Bromo-2-pyridylamine, 6-Bromopyridin-2-amine
A119408-25G
A119408-100G
ALA119408-25GEA
212.71
USD
ALA119408-25G
ALA119408-100G
2-Amino-6-bromopyridine ≥98%
2-Amino-6-bromopyridine
Formula:
C₅H₅BrN₂ MW: 173.01 g/mol Melting Pt: 88-91°C |
MDL Number:
MFCD00137843 CAS Number: 19798-81-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...