2-Amino-6-methoxypyridine ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A119397-100G A119397-1G A119397-25G A119397-5G
ALA119397-100GEA 1842.91 USD
ALA119397-100G ALA119397-1G ALA119397-25G ALA119397-5G
2-Amino-6-methoxypyridine ≥97%
2-Amino-6-methoxypyridine
Formula: C₆H₈N₂O
Boiling Pt: 115°C
Density: 1.15 g/cm³
Flash Pt: 93 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04972542
CAS Number: 17920-35-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e81fe6f8946f8b4fba7414e3dde1486f1375925&vid=6e81fe6f8946f8b4fba7414e3dde1486f1375925&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR