(R)-(-)-3-Aminopiperidine dihydrochloride ≥98%
Supplier: Aladdin Scientific
|
Danger
|
A119349-100G
A119349-1G
A119349-250MG
A119349-25G
A119349-5G
ALA119349-100GEA
5565.96
USD
ALA119349-100G
ALA119349-1G
ALA119349250MG
ALA119349-25G
ALA119349-5G
(R)-(-)-3-Aminopiperidine dihydrochloride ≥98%
(R)-(-)-3-Aminopiperidine dihydrochloride
Formula:
C₅H₁₄Cl₂N₂ Melting Pt: 190-195°C Storage Temperature: Ambient |
MDL Number:
MFCD06799458 CAS Number: 334618-23-4 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...