2-Amino-4,6-dichloropyrimidine-5-carbaldehyde ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

Synonyms: 2-Amino-4,6-dichloropyrimidine-5-carboxaldehyde

A119348-25G A119348-5G A119348-100G A119348-1G
ALA119348-25GEA 1279.56 USD
ALA119348-25G ALA119348-5G ALA119348-100G ALA119348-1G
2-Amino-4,6-dichloropyrimidine-5-carbaldehyde ≥97%
2-Amino-4,6-dichloropyrimidine-5-carbaldehyde
Formula: C₅H₃Cl₂N₃O
MW: 192.00 g/mol
Melting Pt: 208-224°C
Storage Temperature: Refrigerator
MDL Number: MFCD03001242
CAS Number: 5604-46-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d6cd06b517d3b57fc1a19fc5f0cbf48496824c6&vid=9d6cd06b517d3b57fc1a19fc5f0cbf48496824c6&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR