6-Aminoindole ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A119320-1G A119320-250MG
ALA119320-1GEA 72.11 USD
ALA119320-1G ALA119320250MG
6-Aminoindole ≥97%
6-Aminoindole
Formula: C₈H₈N₂
MW: 132.16 g/mol
Boiling Pt: 161-166°C
Melting Pt: 75-79°C
Storage Temperature: Freezer
MDL Number: MFCD02093959
CAS Number: 5318-27-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74f60b8139b68b2ab6722c62cc7851e299e41b9b&vid=74f60b8139b68b2ab6722c62cc7851e299e41b9b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR