2-Amino-5-bromobenzothiazole ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A119305-1G A119305-25G A119305-5G
ALA119305-1GEA 32.21 USD
ALA119305-1G ALA119305-25G ALA119305-5G
2-Amino-5-bromobenzothiazole ≥97%
2-Amino-5-bromobenzothiazole

Product Application:2-Amino-5-bromobenzothiazole is used as pharmaceutical intermediates

Formula: C₇H₅BrN₂S
MW: 229.1 g/mol
Melting Pt: 194-198°C
MDL Number: MFCD00577806
CAS Number: 20358-03-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a75ddb67af9ef635c2786d260e6b2c6ff956175c&vid=a75ddb67af9ef635c2786d260e6b2c6ff956175c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR