Sodium anthraquinone-1-sulfonate ≥98%

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: Anthraquinone-1-sulphonic acid Sodium Salt, 9,10-Dihydro-9,10-dioxo-1-anthraquinonesulphonic acid sodium salt, 1-Anthraquinonesulphonic acid sodium salt, Sodium 9,10-dioxo-9,10-dihydroanthracene-1-sulphonate

A119292-100G A119292-25G A119292-5G
ALA119292-100GEA 130.06 USD
ALA119292-100G ALA119292-25G ALA119292-5G
Sodium anthraquinone-1-sulfonate ≥98%
Sodium anthraquinone-1-sulphonate
Formula: C₁₄H₈O₅S
MW: 310.25 g/mol
MDL Number: MFCD00037145
CAS Number: 128-56-3
EINECS: 204-894-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e761df772fac13eede1a4fe37766776e2fd2e3b9&vid=e761df772fac13eede1a4fe37766776e2fd2e3b9&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR