2-Acetylbenzo[b]thiophene ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 2-Acetylbenzothiophene

A119133-100G A119133-1G A119133-25G A119133-5G
ALA119133-100GEA 1238.71 USD
ALA119133-100G ALA119133-1G ALA119133-25G ALA119133-5G
2-Acetylbenzo[b]thiophene ≥98%
2-Acetylbenzo[b]thiophene

Starter in benzothiophene chemistry.

Formula: C₁₀H₈OS
Melting Pt: 86-90°C
MDL Number: MFCD00090217
CAS Number: 22720-75-8
EINECS: 245-177-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa62665fcd4212644955ce3f1fc3965d17062f6e&vid=fa62665fcd4212644955ce3f1fc3965d17062f6e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR