3-Methylisothiazol-5-amine hydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A119078-1G A119078-25G A119078-5G
ALA119078-1GEA 13.21 USD
ALA119078-1G ALA119078-25G ALA119078-5G
3-Methylisothiazol-5-amine hydrochloride ≥97%
3-Methylisothiazol-5-amine hydrochloride

Application:5-Amino-3-methyl-isothiazole hydrochloride may be used in chemical synthesis studies.Used for pharmacological studies as a precursor for:1.MMP12 inhibitors2.Aurora kinase inhibitors3.N-Heterocyclic indolyl glyoxylamides as anticancer agents.

Formula: C₄H₇ClN₂S
MDL Number: MFCD00012716
CAS Number: 52547-00-9
EINECS: 257-997-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e6c39333a070d631822586d495083ab93d6e075&vid=0e6c39333a070d631822586d495083ab93d6e075&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR