Aluminium oxide ≥99.998% (metals basis)

Supplier: Aladdin Scientific

A119032-100G A119032-25G
ALA119032-100GEA 528.11 USD
ALA119032-100G ALA119032-25G
Aluminium oxide ≥99.998% (metals basis)
Aluminium oxide
Formula: Al₂O₃
Boiling Pt: 2977°C
Melting Pt: 2000°C
Density: 4 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=682ac96f74251817aadd95e7ab503dbbeed64ec2&vid=682ac96f74251817aadd95e7ab503dbbeed64ec2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR