Allylchloro[1,3-bis(2,6-di-i-propylphenyl)imidazol-2-ylidene]palladium(II) ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: Allyl[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]chloropalladium(II), (NHC)Pd(allyl)Cl, Allylchloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]palladium(II), CX 21

A119021-100MG A119021-1G A119021-250MG
ALA119021100MGEA 45.51 USD
ALA119021100MG ALA119021-1G ALA119021250MG
Allylchloro[1,3-bis(2,6-di-i-propylphenyl)imidazol-2-ylidene]palladium(II) ≥98%
Allylchloro[1,3-bis(2,6-di-i-propylphenyl)imidazol-2-ylidene]palladium(II)

The content of this cell is too long for an XLSX file (more than 32767 characters). Please use the CSV format for this export.

Formula: C₃₀H₄₁ClN₂Pd
Melting Pt: >300°C
Storage Temperature: Refrigerator
MDL Number: MFCD08561131
CAS Number: 478980-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR