N-Acetyl-β-D-glucosamine ≥98% for cell culture

Supplier: ALADDIN SCIENTIFIC CORPORATION

Synonyms: 2-Acetamido-2-deoxy-D-glucopyranose, 2-Acetamido-2-deoxy-D-glucose, D-GlcNAc, N-Acetyl-D-glucosamine

A118965-100G A118965-25G A118965-5G
ALA118965-100GEA 226.01 USD
ALA118965-100G ALA118965-25G ALA118965-5G
N-Acetyl-β-D-glucosamine ≥98% for cell culture
N-Acetyl-β-D-glucosamine

N-Acetylglucosamine (GlcNAc), a derivative of glucose is a monosaccharide. It is a white powder and is mildly sweet. It melts at 221°C. GlcNAc is 25% soluble in water. It is present in bromelain, ricin agglutinin and abrus agglutinin, in plants. In humans, it is present in tissue plasminogen activator, growth factors and hormones.

Formula: C₈H₁₅NO₆
MW: 221.21 g/mol
Melting Pt: 211°C
Density: 1 g/cm³
Storage Temperature: Freezer
MDL Number: MFCD00136044
CAS Number: 7512-17-6
EINECS: 231-368-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR