Ac-Ile-Glu-Pro-Asp-PNA ≥97% (by HPLC)

Supplier: ALADDIN SCIENTIFIC

Synonyms: N-Acetyl-Ile-Glu-Pro-Asp-p-nitroanilide, Ac-Ile-Glu-Thr-Asp-pNA, Ac-IETD-pNA

A118745-100MG A118745-25MG A118745-5MG
ALA118745100MGEA 2850.86 USD
ALA118745100MG ALA118745-25MG ALA118745-5MG
Ac-Ile-Glu-Pro-Asp-PNA ≥97% (by HPLC)
Ac-Ile-Glu-Pro-Asp-PNA
Formula: C₂₈H₃₈N₆O₁₁
MW: 634.643 g/mol
Storage Temperature: Freezer
MDL Number: MFCD02259221
CAS Number: 216757-29-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46fbc3ce1e9e341659039fe8a85e1efb7c2bc271&vid=46fbc3ce1e9e341659039fe8a85e1efb7c2bc271&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR