Acetic acid, UltraPureChrom™ for LC-MS
Supplier: ALADDIN SCIENTIFIC
|
Danger
|
Synonyms:
Vinegar acid, Methanecarboxylic acid, Ethanoic acid
A116168-100ML
A116168-25ML
ALA116168-100MLEA
71.9
USD
ALA116168-100ML
ALA116168-25ML
Acetic acid, UltraPureChrom™ for LC-MS
Acetic acid
Formula:
H₃CCOOH MW: 60.05 g/mol Boiling Pt: 118 Melting Pt: 17 Flash Pt: 40°C Storage Temperature: Ambient |
MDL Number:
MFCD00036152 CAS Number: 64-19-7 EINECS: 231-791-2 UN: 2790 ADR: 8 II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...