Ampicillin sodium salt, PharmPure™ USP

Supplier: ALADDIN SCIENTIFIC
Danger

Synonyms: D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt

A105483-25G A105483-100G
ALA105483-25GEA 54.9 USD
ALA105483-25G ALA105483-100G
Ampicillin sodium salt, PharmPure™ USP
Ampicillin sodium salt

Ampicillin is reported as slightly soluble in water, practically insoluble in alcohol, chloroform, ether and fixed oils but soluble in dilute acids or bases. The solution should not be autoclaved; a stock solution should be sterilized through filtration and stored frozen, where it will be stable for months.Used to select for ampicillin resistance in mutated and transformed cells.

Formula: C₁₆H₁₈N₃NaO₄S
MW: 371.39 g/mol
Melting Pt: 215°C
Storage Temperature: Refrigerator
MDL Number: MFCD00064313
CAS Number: 69-52-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbb1bfe6d18837e1f5d4c11624385195dd9bb487&vid=bbb1bfe6d18837e1f5d4c11624385195dd9bb487&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR