Vessel Cannulation Forceps
Supplier: WORLD PRECISION INSTRUMENTS LLC
500453 500377 503375
77662-358EA 160.15 USD
77662-358 77662-328 77663-298
Vessel Cannulation Forceps
Forceps
Vessel cannulation forceps are designed to insert small catheters into vessels.The small hollow jaws of the forceps ensure a secure insertion without damaging the fine tubes.
  • Stainless steel
  • Swiss made