Microintraocular Forceps
Supplier: WORLD PRECISION INSTRUMENTS LLC
501212 500373
77662-384EA 363.41 USD
77662-384 77662-326
Microintraocular Forceps
Forceps
Microintraocular Forceps, also known as microforceps or microsurgical forceps, are specialized surgical instruments used in ophthalmic procedures, particularly in intraocular surgeries.
  • Designed for ophthalmic procedures
  • Surgical grade stainless steel
They have a fine, slender profile and fine tips that allow for precise grasping, manipulation, and dissection of tissues within the eye. They may also be used to manipulate fine sutures, remove fragments or foreign bodies from the eye, or perform intricate tissue dissection.
These intraocular forceps have straight tips. These are made of surgical grade stainless steel, and they are autoclavable.