Liston Bone Cutting Forceps
Supplier: WORLD PRECISION INSTRUMENTS LLC
500358 500462
77662-300EA 113.96 USD
77662-300 77662-372
Liston Bone Cutting Forceps
Forceps
Liston bone cutting forceps, or simply liston forceps, are a surgical instrument for cutting or dividing bones during surgical procedures. Life science researchers might use these robust cutting forceps for gross dissection or tissue dissection, or in plant research to trim stems and roots.
  • Straight tips
  • Spring handles for smooth cutting action
  • Surgical grade stainless steel
Liston forceps have a heavy construction with long handles and sharp, curved blades at the working end. The blades have a concave cutting edge to focus the cutting force and ensures a precise bone cut. These range from 14 cm (5.5") to 19 cm (7.5") long with straight, curved tips.
The longer the handles on the Liston Forceps, the more leverage you have for generating force great enough to cut through bone. The sharp and curved blades enable precise cutting and also reduce the force required to cut through bone. The curved tips help focus the cutting force away fro surrounding tissues, minimizing collateral damage. After making a cut, use your Liston forceps to grasp and remove the bone fragments.
These surgical instruments are made from surgical grade stainless steel and are suitable for sterilization by autoclave or chemical sterilant. Always clean your surgical instruments thoroughly and promptly every time you use them.