Human Recombinant PLGF-3 (from E. coli)
Supplier: FUJIFILM IRVINE SCIENTIFIC, INC
100-198-100UG 100-198-1MG 100-198-25UG 100-198-500UG 100-198-5UG
77660-712EA 607.7 USD
77660-712 77660-714 77660-716 77660-718 77660-720
Human Recombinant PLGF-3 (from E. coli)
Proteins and Peptides
Placenta growth factor (PlGF) is a member of the vascular endothelial growth factor (VEGF) family and promotes endothelial cell growth and angiogenesis. PlGF is secreted as a homodimer and may also form a heterodimer with VEGF. PlGF is detected in the placenta, heart, lungs, thyroid, and adipose tissues. Circulating PlGF levels are correlated with colorectal and renal cancers, as well as atherosclerosis and ischemic heart disease. There are four alternatively spliced PlGF isoforms (PlGF-1, PlGF-2, PlGF-3, and PlGF-4), each with unique secretion patterns and heparin-binding affinities. PlGF-3 lacks a heparin-binding domain and signals through the VEGFR-1 receptor.
  • High quality
  • Low endotoxin
  • Affordable
Precaución: For research use only.