Stevens Needle Holders, Straight
Supplier: WORLD PRECISION INSTRUMENTS LLC
14133 14133-G
77661-790EA 280.28 USD
77661-790 77661-792
Stevens Needle Holders, Straight
Dissecting and Inoculating Loop and Needle Holders
A Stevens needle holder (also called a needle driver) is a surgical instrument used primarily in ophthalmological surgeries to grasp and manipulate surgical needles. A Stevens needle holder has a scissor-like design, with one handle shorter than the other. The shorter handle is held by the surgeon's thumb and index finger, while the longer handle is held with the other fingers.
  • 1 mm serrated jaws
  • Stainless steel or premium German steel
This configuration gives the surgeon a more stable grasp and greater control of the needle holder, which is critical for delicate procedures on small, fragile structures like the eye.
The shorter handle also places the surgeon's hand closer to the surgical site, which reduces the travel distance when manipulating the needle. This helps reduce fatigue and hand tremors. The tips of the jaw are serrated for a firm grasp of the needle for precision and accuracy when manipulating the needle.
Our Stevens needle driver is available in surgical grade stainless steel or our premium German steel. It can be sterilized with a chemical sterilant or by autoclaving. Be sure to clean, disinfect and sterilize your surgical instruments promptly after each use to ensure they last for many years.