Rochester-Carmalt Hemostatic Forceps
Supplier: WORLD PRECISION INSTRUMENTS LLC
501716 501717 501717-G 501243 501243-G 501287
77662-706EA 64.69 USD
77662-706 77662-708 77662-710 77662-478 77662-480 77662-558
Rochester-Carmalt Hemostatic Forceps
Forceps
The Rochester-Carmalt forceps are characterized by the longitudinal serrations that run the length of the blade with cross-hatching at the tip.
  • Longitudinally serrated blade with cross-hatched tip
  • Securely clamp tissue without cutting it
  • Scissor-like ratchet handles
  • Straight or curved tip in a variety of lengths
These large, crushing hemostatic forceps are a choice instrument for clamping blood vessels and large tissues or ligating pedicles. The textured blades, along with the scissor like ratchet handles, ensure a strong grip of the clamped tissue with minimal chance of cutting the tissue during a procedure. One application for the Rochester-Carmalt Forceps is an ovariohysterectomy, where this instrument is used for crushing the vessels of the ovarian pedicle.